Learn More
Invitrogen™ 5-ROX (5-Carboxy-X-Rhodamine, Triethylammonium Salt), single isomer
Description
The carboxylic acid of 5-ROX is used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lisaamine™ rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red™ conjugates.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | 5-ROX |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Carboxylic Acid |
| Label or Dye | ROX |
| Label Type | Classic Dyes |
Frequently Asked Questions (FAQs)
Rhodamine is a generic term for a wide variety of cationic dyes whose fluorescence emission can range from green, orange to red. The table below lists the excitation and emission maxima (nm), as well as molar extinction coefficients (EC; cm-1 M-1), for various rhodamine dyes (data derived with dye dissolved in methanol).
| Dye | Excitation | Emission | EC |
| Rhodamine B | 568 | 583 | 88,000 |
| Rhodamine 123 | 507 | 529 | 101,000 |
| Rhodamine 110 | 499 | 521 | 92,000 |
| Rhodamine 6G | 528 | 551 | 105,000 |
| XRITC | 572 | 596 | 92,000 |
For Research Use Only. Not for use in diagnostic procedures.