missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-endo,3-endo-dimethanol 98.0+%, TCI America™

Numéro de catalogue. N07681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N07681G 1 g
N07685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N07681G Fournisseur TCI America Code fournisseur N07681G
Il en reste null

Identifiants chimiques

CAS 699-97-8
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
InChI Key IGHHPVIMEQGKNE-WZENYGAOSA-N
Synonym 5-norbornene-2-endo,3-endo-dimethanol, norborna-5-ene-2alpha,3alpha-dimethanol, bicyclo 2.2.1 hept-5-ene-2,3-dimethanol, 2r,3s-rel-9ci, 2s,3r-3-hydroxymethyl bicyclo 2.2.1 hept-5-en-2-yl methanol
PubChem CID 12253651
IUPAC Name [(2R,3S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO

Spécifications

Melting Point 86°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2-endo,3-endo-dimethanol