missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentanol 90.0+%, TCI America™

Numéro de catalogue. p-7124625
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C08155ML 5 mL
C081525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C08155ML Fournisseur TCI America Code fournisseur C08155ML
Il en reste null

Identifiants chimiques

CAS 5259-98-3
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.59
MDL Number MFCD00039556
InChI Key DCBJCKDOZLTTDW-UHFFFAOYSA-N
Synonym 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l
PubChem CID 78915
IUPAC Name 5-chloropentan-1-ol
SMILES OCCCCCCl

Spécifications

Color Yellow
Boiling Point 99°C
Quantity 5 mL
Formula Weight 122.59
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-1-pentanol
EINECSNumber (2)-2002
TSCA Yes
Recommended Storage Refrigerator