missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405LS Microplate Washer
GREENER_CHOICE

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.

Supplier:  Agilent BioTek 405LSUSSN

 Afficher plus de versions de ce produit

Numéro de catalogue. BT405LSUS

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. Washing protocols range from gentle cell washing to vigorous ELISA workflows. The 405 LS has a simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols within the extensive onboard software

For use under computer control, Agilent BioTek LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek BioStack is available for walk-away automation of up to 50 microplates.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Certifications

CE Marked

Spécifications

Identifiants chimiques

CE and TUV marked. RoHS compliant.
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Microplate Washer
1 to 250
10 to 85%, noncondensing
1 μL
35.6 cm
25.4 cm
43.2 cm
100/240 V
Optional (4 or 10 L)
25 to 3000 μL/well
<2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
High to Low Flow; Optimized rates for cell assays
50/60 Hz
14 in.
10 in.
17 in.
With internal buffer switching - 36 lb.; Without - 30 lb.

Spécifications

CE and TUV marked. RoHS compliant.
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Microplate Washer
1 to 250
96/384-well Plate Washing
50/60 Hz
1 μL
14 in.
10 in.
17 in.
With internal buffer switching - 36 lb.; Without - 30 lb.
Optional (4 or 10 L)
25 to 3000 μL/well
<2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
High to Low Flow; Optimized rates for cell assays
10 to 85%, noncondensing
405LSUS
Optimized Cell Washing
35.6 cm
25.4 cm
43.2 cm
100/240 V
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions