missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405LS Microplate Washer
GREENER_CHOICE

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.

$17,007.30 - $44,120.36

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Bottles Optional (4 or 10 L)
Precision <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
Dispense Volume 25 to 3000 μL/well
Plate Type 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates
View More Specs

Products 14
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
BT405LSR
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRSN
405LSR Microplate Washer N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRS
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRSSN
405LSRS Ultrasonic Advantage N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRV
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRVSN
405LSRV 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRVS
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRVSSN
405LSRVS 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSHT
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSHTSN
405LSHT Ultrasonic Advantage N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSHTV
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSHTVSN
405LSHTV 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSU
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUSN
405LSU Optimized Cell Washing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSUS
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUSSN
405LSUS Optimized Cell Washing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSUV
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUVSN
405LSUV Optimized Cell Washing, 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSUVS
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUVSSN
405LSUVS Optimized Cell Washing, 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRSQ
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRSQSN
405LSRSQ Ultrasonic Advantage, Verify Technology N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSRVSQ
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSRVSQSN
405LSRVSQ 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSUSQ
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUSQSN
405LSUSQ Optimized Cell Washing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BT405LSUVSQ
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405LSUVSQSN
405LSUVSQ Optimized Cell Washing, 4-Buffer Switching N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. Washing protocols range from gentle cell washing to vigorous ELISA workflows. The 405 LS has a simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols within the extensive onboard software

For use under computer control, Agilent BioTek LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek BioStack is available for walk-away automation of up to 50 microplates.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Specifications

CE and TUV marked. RoHS compliant.
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Microplate Washer
1 to 250
10 to 85%, noncondensing
1 μL
35.6 cm
25.4 cm
43.2 cm
100/240 V
Optional (4 or 10 L)
25 to 3000 μL/well
<2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
High to Low Flow; Optimized rates for cell assays
50/60 Hz
14 in.
10 in.
17 in.
With internal buffer switching - 36 lb.; Without - 30 lb.
Videos
SDS
Documents

Documents