Learn More
Agilent BioTek 405LS Microplate Washer

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.
Supplier: Agilent BioTek 405LSRSSN
Description
The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. Washing protocols range from gentle cell washing to vigorous ELISA workflows. The 405 LS has a simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols within the extensive onboard software
For use under computer control, Agilent BioTek LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek BioStack is available for walk-away automation of up to 50 microplates.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Certifications
CE Marked
Identifiants chimiques
| CE and TUV marked. RoHS compliant. | |
| <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
| 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates | |
| 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
| Microplate Washer | |
| 1 to 250 | |
| 10 to 85%, noncondensing | |
| 1 μL | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| 100/240 V |
| Optional (4 or 10 L) | |
| 25 to 3000 μL/well | |
| <2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells | |
| Programmable in minutes and seconds, up to 60 min. | |
| With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 50/60 Hz | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| With internal buffer switching - 36 lb.; Without - 30 lb. |
Spécifications
| CE and TUV marked. RoHS compliant. | |
| <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
| 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates | |
| 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
| Microplate Washer | |
| 1 to 250 | |
| 96-well Plate Washing | |
| 50/60 Hz | |
| 1 μL | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| With internal buffer switching - 36 lb.; Without - 30 lb. |
| Optional (4 or 10 L) | |
| 25 to 3000 μL/well | |
| <2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells | |
| Programmable in minutes and seconds, up to 60 min. | |
| With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 10 to 85%, noncondensing | |
| 405LSRS | |
| Ultrasonic Advantage | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| 100/240 V |