Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Lift Front Arc Flash Hood
Catalog No. 17001728
Encompass
Change view
Click to view available options
40 Cal ArcGuard RevoLite Lift Front Arc Flash Hood Large
Catalog No.
17001728 40 Cal ArcGuard RevoLite Lift Front Arc Flash Hood Large
1 options
Catalog No. 17001728 Supplier National Safety Apparel™ Supplier No. H65NPQH40LFL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lift front shield makes it easy to obtain fresh air while out of hazard zone
  • Knit gussets at side provide greater range of motion and flexibility
  • Premium anti-fog and scratch resistant coating on face shield
  • PureView face shield technology features a greater range of visibility
  • Hard hat included with slotted adapted
  • Vents provide fresh air and increased auditory capabilities
Specifications
Size Medium
Material RevoLite™
Color Olive Green
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269