Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite Crossvent Hood with PureView Faceshield
Catalog No. 17001729
Encompass
Change view
Click to view available options
Color: Olive green
Catalog No.
17001729 Color: Olive green
1 options
Catalog No. 17001729 Supplier National Safety Apparel™ Supplier No. H65NPQHFANPV
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Technology which features a single layer lens & greater range of visibility. Premium anti-fog & scratch-resistant coating on faceshield
  • Fans are attached with FR hook & loop
  • Fans are powered by 9-volt batteries
  • FR fabric that will NOT melt, drip, or ignite
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog & scratch resistant coating on faceshield
  • Inherently flame resistant
  • Hard hat included
Specifications
Size Medium
Material RevoLite™
Color Olive Green
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269