Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance Lift Front Arc Flash Hood
Catalog No. 17001734
Encompass
Change view
Click to view available options
Color: Navy
Catalog No.
17001734 Color: Navy
1 options
Catalog No. 17001734 Supplier National Safety Apparel™ Supplier No. H65UQUQ40LF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Has a lift front hood with PureView™ faceshield technology for quick access to fresh air and a clearer view
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • Knit gussets at side provide greater range of motion and flexibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • PureView faceshield technology features a greater range of visibility
  • Hard hat included
  • Vents provide fresh air and increased auditory capabilities
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269