missing translation for 'onlineSavingsMsg'
Learn More

[4-(Tetrahydropyran-4-yloxy)phenyl]methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC52413DA

Quantity 1g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC52413DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

864266-61-5
207.273
QPNQGXQLNABWSW-UHFFFAOYSA-N
24229525
C1COCCC1OC2=CC=C(C=C2)CN
C12H17NO2
MFCD09064967
4-tetrahydropyran-4-yloxy phenyl methylamine, 4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy benzylamine, 4-4-aminomethyl phenoxy tetrahydro-2h-pyran, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methylamine, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methylamine, benzenemethanamine,4-tetrahydro-2h-pyran-4-yl oxy
[4-(oxan-4-yloxy)phenyl]methanamine

Spécifications

[4-(Tetrahydropyran-4-yloxy)phenyl]methylamine
96.5
97%
C12H17NO2
MFCD09064967
QPNQGXQLNABWSW-UHFFFAOYSA-N
[4-(oxan-4-yloxy)phenyl]methanamine
24229525
97%
864266-61-5
100.0
Amber Glass Bottle
1g
4-tetrahydropyran-4-yloxy phenyl methylamine, 4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy benzylamine, 4-4-aminomethyl phenoxy tetrahydro-2h-pyran, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methylamine, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methylamine, benzenemethanamine,4-tetrahydro-2h-pyran-4-yl oxy
C1COCCC1OC2=CC=C(C=C2)CN
207.273
207.27
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS
Documents

Documents