missing translation for 'onlineSavingsMsg'
Learn More

[4-(Tetrahydropyran-4-yloxy)phenyl]methylamine, 97%, Thermo Scientific™

Catalog No. p-3830156
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC52413CB 250mg Amber Glass Bottle
CC52413DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC52413CB Supplier Thermo Scientific Chemicals Supplier No. CC52413CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864266-61-5
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD09064967
InChI Key QPNQGXQLNABWSW-UHFFFAOYSA-N
Synonym 4-tetrahydropyran-4-yloxy phenyl methylamine, 4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yloxy phenyl methanamine, 1-4-oxan-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy benzylamine, 4-4-aminomethyl phenoxy tetrahydro-2h-pyran, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, 4-tetrahydro-2h-pyran-4-yl oxy phenyl methylamine, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methylamine, benzenemethanamine,4-tetrahydro-2h-pyran-4-yl oxy
PubChem CID 24229525
IUPAC Name [4-(oxan-4-yloxy)phenyl]methanamine
SMILES C1COCCC1OC2=CC=C(C=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material [4-(Tetrahydropyran-4-yloxy)phenyl]methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.