missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobiphenyl 97.0+%, TCI America™

Catalog No. F04825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04825G 5 g
F04821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04825G Supplier TCI America Supplier No. F04825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 324-74-3
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.202
MDL Number MFCD00011650
InChI Key RUYZJEIKQYLEGZ-UHFFFAOYSA-N
Synonym 4-fluorobiphenyl, 4-fluoro-1,1'-biphenyl, p-fluorodiphenyl, 4-fluoro-biphenyl, 4-fluorodiphenyl, biphenyl, 4-fluoro, 1,1'-biphenyl, 4-fluoro, para-fluorobiphenyl, 4-fluorophenyl benzene, p-fluorobiphenyl
PubChem CID 9461
IUPAC Name 1-fluoro-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)F

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 253°C
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobiphenyl
RTECSNumber DV5291200
TSCA No