missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobiphenyl, 97+%

Catalog No. AAL0717006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0717006 5 g
AAL0717003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0717006 Supplier Thermo Scientific Chemicals Supplier No. L0717006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 324-74-3 | C12H9F | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 324-74-3
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.202
MDL Number MFCD00011650
InChI Key RUYZJEIKQYLEGZ-UHFFFAOYSA-N
Synonym 4-fluorobiphenyl, 4-fluoro-1,1'-biphenyl, p-fluorodiphenyl, 4-fluoro-biphenyl, 4-fluorodiphenyl, biphenyl, 4-fluoro, 1,1'-biphenyl, 4-fluoro, para-fluorobiphenyl, 4-fluorophenyl benzene, p-fluorobiphenyl
PubChem CID 9461
IUPAC Name 1-fluoro-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)F

Specifications

Melting Point 72°C to 76°C
Boiling Point 253°C
Quantity 5 g
Beilstein 2042410
Formula Weight 172.2
Percent Purity ≥97%
Chemical Name or Material 4-Fluorobiphenyl
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-304-1
RTECSNumber DV5291200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only