missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylbenzenesulfonamide 98.0+%, TCI America™
$84.70 - $256.05
Chemical Identifiers
| CAS | 138-38-5 |
|---|---|
| Molecular Formula | C8H11NO2S |
| Molecular Weight (g/mol) | 185.24 |
| MDL Number | MFCD01365828 |
| InChI Key | MLTGAVXHWSDGIS-UHFFFAOYSA-N |
| PubChem CID | 222870 |
| IUPAC Name | 4-ethylbenzene-1-sulfonamide |
| SMILES | CCC1=CC=C(C=C1)S(N)(=O)=O |
Chemical Identifiers
| 138-38-5 | |
| 185.24 | |
| MLTGAVXHWSDGIS-UHFFFAOYSA-N | |
| 4-ethylbenzene-1-sulfonamide |
| C8H11NO2S | |
| MFCD01365828 | |
| 222870 | |
| CCC1=CC=C(C=C1)S(N)(=O)=O |
Specifications
| 138-38-5 | |
| White-Yellow | |
| MFCD01365828 | |
| MLTGAVXHWSDGIS-UHFFFAOYSA-N | |
| 4-ethylbenzene-1-sulfonamide | |
| 222870 | |
| ≥98.0% (HPLC,N) | |
| 4-Ethylbenzenesulfonamide |
| 111°C | |
| C8H11NO2S | |
| 5 g | |
| CCC1=CC=C(C=C1)S(N)(=O)=O | |
| 185.24 | |
| 185.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0233
TSCA : No