missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylbenzenesulfonamide 98.0+%, TCI America™
Supplier: TCI America E077325G
Specifications
| 4-Ethylbenzenesulfonamide | |
| 111°C | |
| C8H11NO2S | |
| 25 g | |
| CCC1=CC=C(C=C1)S(N)(=O)=O | |
| 185.24 | |
| 185.24 | |
| Crystalline Powder |
| 138-38-5 | |
| White-Yellow | |
| MFCD01365828 | |
| MLTGAVXHWSDGIS-UHFFFAOYSA-N | |
| 4-ethylbenzene-1-sulfonamide | |
| 222870 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 138-38-5 | |
| 185.24 | |
| MLTGAVXHWSDGIS-UHFFFAOYSA-N | |
| 4-ethylbenzene-1-sulfonamide |
| C8H11NO2S | |
| MFCD01365828 | |
| 222870 | |
| CCC1=CC=C(C=C1)S(N)(=O)=O |
Safety and Handling
EINECSNumber : (9)-0233
TSCA : No