missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzenesulfonamide 98.0+%, TCI America™

Supplier:  TCI America E07735G

 View more versions of this product

Catalog No. E07735G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Ethylbenzenesulfonamide
111°C
C8H11NO2S
5 g
CCC1=CC=C(C=C1)S(N)(=O)=O
185.24
185.24
Crystalline Powder
138-38-5
White-Yellow
MFCD01365828
MLTGAVXHWSDGIS-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonamide
222870
≥98.0% (HPLC,N)

Chemical Identifiers

138-38-5
185.24
MLTGAVXHWSDGIS-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonamide
C8H11NO2S
MFCD01365828
222870
CCC1=CC=C(C=C1)S(N)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0233

TSCA : No

SDS
Documents

Documents

Promotions

Promotions