accessibility menu, dialog, popup

UserName

H3CSNH2OO

CAS RN 138-38-5

IUPAC Name: 4-ethylbenzene-1-sulfonamide
Synonyms: 4-ethylbenzenesulfonamide
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: MLTGAVXHWSDGIS-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results