missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline, 98+%

CAS: 38762-41-3 | C6H5BrClN | 206.467 g/mol

$81.56 - $925.01

Chemical Identifiers

CAS 38762-41-3
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00007660
InChI Key INMZDDDQLHKGPF-UHFFFAOYSA-N
Synonym 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
PubChem CID 610169
IUPAC Name 4-bromo-2-chloroaniline
SMILES C1=CC(=C(C=C1Br)Cl)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1785909
View Documents
Thermo Scientific Chemicals
A1785909
10 g
Each for $81.56
Only null left
 
AAA1785918
View Documents
Thermo Scientific Chemicals
A1785918
50 g
Each for $263.03
Only null left
 
AAA1785930
View Documents
Thermo Scientific Chemicals
A1785930
250 g
Each for $925.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38762-41-3
206.467
INMZDDDQLHKGPF-UHFFFAOYSA-N
610169
C1=CC(=C(C=C1Br)Cl)N
C6H5BrClN
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
4-bromo-2-chloroaniline

Specifications

38762-41-3
C6H5BrClN
10 g
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
C1=CC(=C(C=C1Br)Cl)N
206.467
206.47
4-Bromo-2-chloroaniline
68°C to 72°C
MFCD00007660
3117740
INMZDDDQLHKGPF-UHFFFAOYSA-N
4-bromo-2-chloroaniline
610169
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 254-118-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only