missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline, 98+%

CAS: 38762-41-3 | C6H5BrClN | 206.467 g/mol

Supplier:  Thermo Scientific Chemicals A1785930

 View more versions of this product

Catalog No. AAA1785930


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-2-chloroaniline
68°C to 72°C
MFCD00007660
3117740
INMZDDDQLHKGPF-UHFFFAOYSA-N
4-bromo-2-chloroaniline
610169
≥98%
38762-41-3
C6H5BrClN
250 g
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
C1=CC(=C(C=C1Br)Cl)N
206.467
206.47

Chemical Identifiers

38762-41-3
206.467
INMZDDDQLHKGPF-UHFFFAOYSA-N
610169
C1=CC(=C(C=C1Br)Cl)N
C6H5BrClN
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
4-bromo-2-chloroaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 254-118-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only