Learn More
4-Bromo-2-chloroaniline, 98+%
CAS: 38762-41-3 | C6H5BrClN | 206.467 g/mol
Supplier: Thermo Scientific Chemicals A1785918
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Bromo-2-chloroaniline | |
| 68°C to 72°C | |
| MFCD00007660 | |
| 3117740 | |
| INMZDDDQLHKGPF-UHFFFAOYSA-N | |
| 4-bromo-2-chloroaniline | |
| 610169 | |
| ≥98% |
| 38762-41-3 | |
| C6H5BrClN | |
| 50 g | |
| 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline | |
| C1=CC(=C(C=C1Br)Cl)N | |
| 206.467 | |
| 206.47 |
Chemical Identifiers
| 38762-41-3 | |
| 206.467 | |
| INMZDDDQLHKGPF-UHFFFAOYSA-N | |
| 610169 | |
| C1=CC(=C(C=C1Br)Cl)N |
| C6H5BrClN | |
| MFCD00007660 | |
| 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline | |
| 4-bromo-2-chloroaniline |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 254-118-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only