missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline, 98+%

Catalog No. aaa1785909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1785909 10 g
AAA1785918 50 g
AAA1785930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1785909 Supplier Thermo Scientific Chemicals Supplier No. A1785909
Only null left

CAS: 38762-41-3 | C6H5BrClN | 206.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38762-41-3
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00007660
InChI Key INMZDDDQLHKGPF-UHFFFAOYSA-N
Synonym 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
PubChem CID 610169
IUPAC Name 4-bromo-2-chloroaniline
SMILES C1=CC(=C(C=C1Br)Cl)N

Specifications

Melting Point 68°C to 72°C
Quantity 10 g
Beilstein 3117740
Formula Weight 206.47
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-chloroaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 254-118-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only