missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-methyl-1-butanol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 103.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Amino-2-methyl-1-butanol |
Sécurité et manipulation
| EINECSNumber | (2)-3927 |
| TSCA | No |