Learn More
4-[(5-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 364793-76-0 |
|---|---|
| Molecular Formula | C9H12BrNOS |
| Molecular Weight (g/mol) | 262.165 |
| MDL Number | MFCD11506356 |
| InChI Key | XTTAOCHKCUOJFW-UHFFFAOYSA-N |
| Synonym | 4-5-bromothiophen-2-yl methyl morpholine, 4-5-bromothien-2-yl methyl morpholine, 5-bromo-2-morpholin-4-ylmethyl thiophene, morpholine,4-5-bromo-2-thienyl methyl, 4-5-bromothien-2-ylmethyl morpholine, 2-bromo-5-morpholin-4-ylmethyl thiophene, 4-5-bromo-thiophen-2-ylmethyl-morpholine |
| PubChem CID | 21914272 |
| IUPAC Name | 4-[(5-bromothiophen-2-yl)methyl]morpholine |
| SMILES | C1COCCN1CC2=CC=C(S2)Br |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|
CC73410DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
CC73410DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 364793-76-0 | |
| 262.165 | |
| XTTAOCHKCUOJFW-UHFFFAOYSA-N | |
| 21914272 | |
| C1COCCN1CC2=CC=C(S2)Br |
| C9H12BrNOS | |
| MFCD11506356 | |
| 4-5-bromothiophen-2-yl methyl morpholine, 4-5-bromothien-2-yl methyl morpholine, 5-bromo-2-morpholin-4-ylmethyl thiophene, morpholine,4-5-bromo-2-thienyl methyl, 4-5-bromothien-2-ylmethyl morpholine, 2-bromo-5-morpholin-4-ylmethyl thiophene, 4-5-bromo-thiophen-2-ylmethyl-morpholine | |
| 4-[(5-bromothiophen-2-yl)methyl]morpholine |
Spécifications
| 364793-76-0 | |
| 100.0 | |
| C9H12BrNOS | |
| 4-5-bromothiophen-2-yl methyl morpholine, 4-5-bromothien-2-yl methyl morpholine, 5-bromo-2-morpholin-4-ylmethyl thiophene, morpholine,4-5-bromo-2-thienyl methyl, 4-5-bromothien-2-ylmethyl morpholine, 2-bromo-5-morpholin-4-ylmethyl thiophene, 4-5-bromo-thiophen-2-ylmethyl-morpholine | |
| C1COCCN1CC2=CC=C(S2)Br | |
| 262.165 | |
| 262.17 | |
| 4-[(5-Bromothien-2-yl)methyl]morpholine |
| 96.5 | |
| 97% | |
| MFCD11506356 | |
| XTTAOCHKCUOJFW-UHFFFAOYSA-N | |
| 4-[(5-bromothiophen-2-yl)methyl]morpholine | |
| 21914272 | |
| 97% |
Sécurité et manipulation
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger