missing translation for 'onlineSavingsMsg'
Learn More

4-[(5-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC73410DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC73410DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

364793-76-0
262.165
XTTAOCHKCUOJFW-UHFFFAOYSA-N
21914272
C1COCCN1CC2=CC=C(S2)Br
C9H12BrNOS
MFCD11506356
4-5-bromothiophen-2-yl methyl morpholine, 4-5-bromothien-2-yl methyl morpholine, 5-bromo-2-morpholin-4-ylmethyl thiophene, morpholine,4-5-bromo-2-thienyl methyl, 4-5-bromothien-2-ylmethyl morpholine, 2-bromo-5-morpholin-4-ylmethyl thiophene, 4-5-bromo-thiophen-2-ylmethyl-morpholine
4-[(5-bromothiophen-2-yl)methyl]morpholine

Specifications

4-[(5-Bromothien-2-yl)methyl]morpholine
96.5
97%
C9H12BrNOS
MFCD11506356
XTTAOCHKCUOJFW-UHFFFAOYSA-N
4-[(5-bromothiophen-2-yl)methyl]morpholine
21914272
97%
364793-76-0
100.0
Amber Glass Bottle
5g
4-5-bromothiophen-2-yl methyl morpholine, 4-5-bromothien-2-yl methyl morpholine, 5-bromo-2-morpholin-4-ylmethyl thiophene, morpholine,4-5-bromo-2-thienyl methyl, 4-5-bromothien-2-ylmethyl morpholine, 2-bromo-5-morpholin-4-ylmethyl thiophene, 4-5-bromo-thiophen-2-ylmethyl-morpholine
C1COCCN1CC2=CC=C(S2)Br
262.165
262.17
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents