missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobiphenyl, TCI America™

Catalog No. D21811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21811G Supplier TCI America Supplier No. D21811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 398-23-2
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.193
MDL Number MFCD00000349
InChI Key PZDAAZQDQJGXSW-UHFFFAOYSA-N
Synonym 4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro
PubChem CID 9811
IUPAC Name 1-fluoro-4-(4-fluorophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)F

Specifications

Melting Point 89°C
Boiling Point 255°C
Quantity 1 g
UN Number 3152
Formula Weight 190.19
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Difluorobiphenyl