missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobiphenyl, 99%

Catalog No. AAL0836106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0836106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0836106 Supplier Thermo Scientific Chemicals Supplier No. L0836106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 398-23-2 | C12H8F2 | 190.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 398-23-2
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.193
MDL Number MFCD00000349
InChI Key PZDAAZQDQJGXSW-UHFFFAOYSA-N
Synonym 4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro
PubChem CID 9811
IUPAC Name 1-fluoro-4-(4-fluorophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)F

Specifications

Melting Point 87°C to 89°C
Boiling Point 254°C to 255°C
Quantity 5 g
UN Number UN3152
Beilstein 2044699
Merck Index 14,3149
Formula Weight 190.19
Percent Purity 99%
Chemical Name or Material 4,4'-Difluorobiphenyl
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
EINECSNumber 206-910-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only