missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ M-300 Series PAPR Replacement Head Suspension

Catalog No. 19039518
Change view
Click to view available options
For Use With (Equipment):
M-300 Series Hardhats and M-400 Series Helmets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19039518 M-300 Series Hardhats and M-400 Series Helmets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19039518 Supplier 3M™ Supplier No. 7000127679

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Head suspension is a replacement part for Versaflo™ M-300 Series hardhats and M-400 Series helmets.

  • 3M Catalog# M-350

Specifications

Color Black
Disposable Reusable
Product Type Head Suspension
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-300 Series Hardhats and M-400 Series Helmets
Model M-350