missing translation for 'onlineSavingsMsg'
Learn More

3M™ Fit Test Solution

Catalog No. 19002722
Change view
Click to view available options
Quantity:
6/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19002722 6/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19002722 Supplier 3M™ Supplier No. 7100350838
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Fit Test Solution used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.

  • Bitter taste fit test solution for fit testing.
  • Used with the 3M™ Qualitative Fit Test Apparatus FT-30.
  • Can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.

Specifications

Product Type Fit Test Solution
Capacity (English) 1.9 oz.
Capacity (Metric) 55 mL
Certifications/Compliance OSHA 29 CFR 1910.134, Appendix A
For Use With (Application) Respirator Fit Testing
For Use With (Equipment) Bitter Qualitative Fit Test Kit
Model FT-32
Quantity 6/Cs.

For Occupational Use Only