missing translation for 'onlineSavingsMsg'
Learn More

3M™ Sensitivity Solution FT-31, Bitter

Catalog No. 19002721
Change view
Click to view available options
Quantity:
6/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19002721 6/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death
Catalog No. 19002721 Supplier 3M™ Supplier No. 7100004635
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death

Bitter sensitivity solution is used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.

 

  • Bitter taste sensitivity solution for fit testing
  • Used with the 3M™ Qualitative Fit Test Apparatus FT-30
  • For occupational use only
  • Used to test the sensitivity of the respirator wearer to a bitter solution. It is used with the 3M™ Qualitative Fit Test Apparatus FT 30

 

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product Misuse may result in sickness or death

Specifications

Product Type Sensitivity Solution
Capacity (English) 1.9 oz.
Capacity (Metric) 55 mL
Certifications/Compliance OSHA 29 CFR 1910.134, Appendix A
For Use With (Application) Respirator Fit Testing
For Use With (Equipment) Bitter Qualitative Fit Test Kit
Model FT-31
Quantity 6/Cs.
Type (Solution) Sensitivity test solution