Learn More
3M™ E-A-Rflex™ 28 Banded Hearing Protector
Catalog No. 296363004
Change view
Click to view available options
Noise Reduction Rating (NRR):
28 dB
Catalog No. Noise Reduction Rating (NRR)
296363004 28 dB
1 options
Catalog No. 296363004 Supplier 3M™ Supplier No. 7000127170
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A comfortable, effective, lightweight alternative to ear muffs E-A-Rflex™ helps provide excellent noise reduction. Great for people who fit and remove their hearing protection frequently. Soft tips pivot to help maintain a comfortable, noise-blocking seal. Band may be worn in multiple positions.
  • Pivoting tips for proper, comfortable fit that moves with the wearer for more consistent protection.
  • Features soft, E-A-Rform™ foam tips that reacts to body temperature for enhanced comfort and fit.
  • Replacement Pods also available.
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 320-1000
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 28 dB
Plug Shape Cone
Disposable Reusable
Metal Detectable No
Dielectric No
Show More Show Less