Learn More
3M™ E-A-R™ Caboflex™ Hearing Protector
Catalog No. 173808
Change view
Click to view available options
CSA Z94.2:
Class AL
Catalog No. CSA Z94.2
173808 Class AL
1 options
Catalog No. 173808 Supplier 3M™ Supplier No. 7000002297
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E-A-R™ Caboflex™ banded hearing protector is lightweight when moving in and out of high noise areas and also fits around the neck when not in use. Each cone-shaped tip pivots to align with the ear, providing both comfort and protection.
  • Silicone tips contain a foam core for an effective seal and good noise reduction.
  • Flexible band can be worn under the chin for use with hard hats.
  • Ideal for intermittent hearing protection needs.
  • Replacement Pods also available.
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility No
Model 320-2001
Pairs per Package 100 Pairs/Pk.
Plug Material SiliCone, PVC
Plug Color Yellow
Noise Reduction Rating (NRR) 17 dB (behind head), 20 dB (under chin)
Plug Shape Cone
Disposable Reusable
Metal Detectable No
Dielectric No
Show More Show Less