Learn More
3M™ Accessories and Replacement Parts for Full-Facepiece Respirators, Inhalation valve
Catalog No. 189994561
Change view
Click to view available options
For Use With (Equipment):
6000 series full facepiece respirator and half mask respirator
Catalog No. For Use With (Equipment)
189994561 6000 series full facepiece respirator and half mask respirator
1 options
Catalog No. 189994561 Supplier 3M™ Supplier No. 7000002049
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Inhalation valve for use with full-facepiece respirators.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# 6893
Specifications
Product Type Inhalation Valve
For Use With (Application) Automotive, metal production, fabrication, pharmaceutical, transportation, military maintenance, construction, food and beverage manufacturing, grinding, sanding, welding
For Use With (Equipment) 6000 series full facepiece respirator and half mask respirator
Model 6893