missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 / 7800 Series Full Facepiece Accessory, Inhalation Port Gasket

Catalog No. 189994562
Change view
Click to view available options
For Use With (Equipment):
6000, 7800 Series Full Facepiece
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
189994562 6000, 7800 Series Full Facepiece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189994562 Supplier 3M™ Supplier No. 7000002050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhalation port gasket for use with full-facepiece respirators.

  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# 6895

Specifications

Color Orange
Product Type Gasket
For Use With (Application) Automotive, metal production, fabrication, pharmaceutical, transportation, military maintenance, construction, food and beverage manufacturing, grinding, sanding, welding
For Use With (Equipment) 6000, 7800 Series Full Facepiece
Model 6895