missing translation for 'onlineSavingsMsg'
Learn More

3M™ Exhalation Valve Replacement Part, 6500 Series Half Mask Respirator

Catalog No. 19096910
Change view
Click to view available options
For Use With (Equipment):
Reusable Half Masks 6500 Series
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19096910 Reusable Half Masks 6500 Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19096910 Supplier 3M™ Supplier No. 7000128253

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Spare part for use with 3M™ 6500 half mask respirator.

  • Enhanced comfort by reducing heat and moisture
  • Easier breathing with unique valve design
  • Replacement part for the 3M™ 6500 Series
  • 10 each per case
  • For occupational use only

Specifications

Connection Type Bayonet
Color White
Product Type Exhalation Valve
For Use With (Equipment) Reusable Half Masks 6500 Series
Model 6583