missing translation for 'onlineSavingsMsg'
Learn More

(3-Phenyl-1,2,4-oxadiazol-5-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC76913DE

Quantity 5g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC76913DE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

90564-77-5
175.191
QFBMJBDECSEYCZ-UHFFFAOYSA-N
4894507
C1=CC=C(C=C1)C2=NOC(=N2)CN
C9H9N3O
MFCD06166274
3-phenyl-1,2,4-oxadiazol-5-yl methanamine, 3-phenyl-1,2,4-oxadiazol-5-yl methylamine, 1-3-phenyl-1,2,4-oxadiazol-5-yl methanamine, c-3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, cbi-bb zero/004833, 5-aminomethyl-3-phenyl-1,2,4-oxadiazole, 3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, 3-phenyl-1,2,4-oxadiazol-5-yl methyl amine
(3-phenyl-1,2,4-oxadiazol-5-yl)methanamine

Spécifications

(3-Phenyl-1, 2, 4-oxadiazol-5-yl)methylamine
96.5
97%
C9H9N3O
MFCD06166274
QFBMJBDECSEYCZ-UHFFFAOYSA-N
(3-phenyl-1,2,4-oxadiazol-5-yl)methanamine
4894507
97%
90564-77-5
100.0
Amber Glass Bottle
5g
3-phenyl-1,2,4-oxadiazol-5-yl methanamine, 3-phenyl-1,2,4-oxadiazol-5-yl methylamine, 1-3-phenyl-1,2,4-oxadiazol-5-yl methanamine, c-3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, cbi-bb zero/004833, 5-aminomethyl-3-phenyl-1,2,4-oxadiazole, 3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, 3-phenyl-1,2,4-oxadiazol-5-yl methyl amine
C1=CC=C(C=C1)C2=NOC(=N2)CN
175.191
175.19
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS
Documents

Documents