missing translation for 'onlineSavingsMsg'
Learn More

(3-Phenyl-1,2,4-oxadiazol-5-yl)methylamine, 97%, Thermo Scientific™

Catalog No. p-3881926
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC76913DA 1g Amber Glass Bottle
CC76913DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC76913DA Supplier Thermo Scientific Chemicals Supplier No. CC76913DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90564-77-5
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD06166274
InChI Key QFBMJBDECSEYCZ-UHFFFAOYSA-N
Synonym 3-phenyl-1,2,4-oxadiazol-5-yl methanamine, 3-phenyl-1,2,4-oxadiazol-5-yl methylamine, 1-3-phenyl-1,2,4-oxadiazol-5-yl methanamine, c-3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, cbi-bb zero/004833, 5-aminomethyl-3-phenyl-1,2,4-oxadiazole, 3-phenyl-1,2,4 oxadiazol-5-yl-methylamine, 3-phenyl-1,2,4-oxadiazol-5-yl methyl amine
PubChem CID 4894507
IUPAC Name (3-phenyl-1,2,4-oxadiazol-5-yl)methanamine
SMILES C1=CC=C(C=C1)C2=NOC(=N2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material (3-Phenyl-1, 2, 4-oxadiazol-5-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.