Learn More
(3-Mercaptopropyl)methyldimethoxysilane, 95%
Description
(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Boiling Point | 96°C (30mmHg) |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Linear Formula | HSCH2CH2CH2Si(OCH3)2CH3 |
| Flash Point | 93°C (199°F) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 180.34 |
| Percent Purity | 95% |
| Refractive Index | 1.45 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H227-H302-H317 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c |
| EINECSNumber | 250-426-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only