missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocyclohexene 94.0+%, TCI America™

Catalog No. B09435ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09435ML 5 mL
B094325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09435ML Supplier TCI America Supplier No. B09435ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1521-51-3
Molecular Formula C6H9Br
Molecular Weight (g/mol) 161.042
MDL Number MFCD00013775
InChI Key AJKDUJRRWLQXHM-UHFFFAOYSA-N
Synonym 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
PubChem CID 137057
IUPAC Name 3-bromocyclohexene
SMILES C1CC=CC(C1)Br

Specifications

Boiling Point 82°C
Quantity 5 mL
UN Number 1993
Formula Weight 161.04
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromocyclohexene
TSCA No
Recommended Storage Refrigerator