missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. D45345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45345G 5 g
D453425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45345G Supplier TCI America Supplier No. D45345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134412-17-2
Molecular Formula C23H28F2
Molecular Weight (g/mol) 342.47
MDL Number MFCD09839000
InChI Key NQLHGECGZRJQLF-UHFFFAOYSA-N
Synonym 3,4-difluoro-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-pentylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-pentylcyclohexyl phenyl benzene, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl, 1,1'-biphenyl,3,4-difluoro-4'-trans-4-pentylcyclohexyl, 3,4-difluoro-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl, acmc-20mvco, 3,4-difluoro-4/'-4-pentylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-pentylcyclohexyl benzene
PubChem CID 612307
IUPAC Name 3,4-difluoro-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
SMILES CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 342.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluoro-4′-(trans-4-pentylcyclohexyl)biphenyl