missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. T32601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32601G 1 g
T32605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32601G Supplier TCI America Supplier No. T32601G

Chemical Identifiers

CAS 132123-39-8
Molecular Formula C21H23F3
Molecular Weight (g/mol) 332.41
MDL Number MFCD09839001
InChI Key RRKRBRVTKIRLHH-UHFFFAOYSA-N
Synonym 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl
PubChem CID 15014921
IUPAC Name 1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F

Specifications

Melting Point 42°C
Color White
Quantity 1 g
Formula Weight 332.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluoro-4′-(trans-4-propylcyclohexyl)biphenyl
EINECSNumber (4)-1980
TSCA No