Learn More
National Safety Apparel™ 20 Cal ArcGuard™ Lift Front Shroud
Catalog No. 17001741
Encompass
Change view
Click to view available options
Color: Navy
Catalog No.
17001741 Color: Navy
1 options
Catalog No. 17001741 Supplier National Safety Apparel™ Supplier No. H65UW20LFS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lift front shroud
  • Arc Rating: 20 cal/cm²
  • CAT 2
  • Fabric/Material: 13 oz. Westex™ UltraSoft™
  • PureView™ faceshield technology features a greater range of visibility
  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • Premium anti-fog and scratch resistant coating on faceshield
  • PureView™ technology for proven color recognition
  • Increased field of view
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Arc Flash Rating 20 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, NFPA 70 E, ASTM F2178, CSA Z462, ANSI/ISEA Z87.1(Impact Rating Z81+)