missing translation for 'onlineSavingsMsg'
Learn More

(2-Thiomorpholinopyrid-4-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC50509DA

Quantity 1g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC50509DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

898289-25-3
210.295
KDJBEJVGUFHDPW-UHFFFAOYSA-N
24229508
C1CSCCN1C2=NC=CC(=C2)CO
C10H14N2OS
MFCD09702362
2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
(2-thiomorpholin-4-ylpyridin-4-yl)methanol

Spécifications

(2-Thiomorpholinopyrid-4-yl)methanol
96.5
97%
C10H14N2OS
MFCD09702362
KDJBEJVGUFHDPW-UHFFFAOYSA-N
(2-thiomorpholin-4-ylpyridin-4-yl)methanol
24229508
97%
898289-25-3
100.0
Amber Glass Bottle
1g
2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
C1CSCCN1C2=NC=CC(=C2)CO
210.295
210.3
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

FDS
Documents

Documents