missing translation for 'onlineSavingsMsg'
Learn More

(2-Thiomorpholinopyrid-4-yl)methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 898289-25-3
Molecular Formula C10H14N2OS
Molecular Weight (g/mol) 210.295
MDL Number MFCD09702362
InChI Key KDJBEJVGUFHDPW-UHFFFAOYSA-N
Synonym 2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
PubChem CID 24229508
IUPAC Name (2-thiomorpholin-4-ylpyridin-4-yl)methanol
SMILES C1CSCCN1C2=NC=CC(=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC50509CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50509DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

898289-25-3
210.295
KDJBEJVGUFHDPW-UHFFFAOYSA-N
24229508
C1CSCCN1C2=NC=CC(=C2)CO
C10H14N2OS
MFCD09702362
2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
(2-thiomorpholin-4-ylpyridin-4-yl)methanol

Specifications

898289-25-3
100.0
C10H14N2OS
2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
C1CSCCN1C2=NC=CC(=C2)CO
210.295
210.3
(2-Thiomorpholinopyrid-4-yl)methanol
96.5
97%
MFCD09702362
KDJBEJVGUFHDPW-UHFFFAOYSA-N
(2-thiomorpholin-4-ylpyridin-4-yl)methanol
24229508
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents