missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Cinnamate 99.0+%, TCI America™

Numéro de catalogue. P200725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P200725G 25 g
P2007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P200725G Fournisseur TCI America Code fournisseur P200725G
Il en reste null

Identifiants chimiques

CAS 103-53-7
Molecular Formula C17H16O2
Molecular Weight (g/mol) 252.313
MDL Number MFCD00022050
InChI Key MJQVZIANGRDJBT-VAWYXSNFSA-N
Synonym Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester
PubChem CID 5369459
IUPAC Name 2-phenylethyl (E)-3-phenylprop-2-enoate
SMILES C1=CC=C(C=C1)CCOC(=O)C=CC2=CC=CC=C2

Spécifications

Melting Point 57°C
Color White-Yellow
Quantity 25 g
Formula Weight 252.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylethyl Cinnamate
EINECSNumber (3)-1739
RTECSNumber GE0405000
TSCA Yes