missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stabilized

Catalog No. AC164965000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164965000 500 mL Glass bottle
AC164961000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164965000 Supplier Thermo Scientific Chemicals Supplier No. 164965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-30-9 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-30-9 , 7440-50-8
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -90.0°C
Color Colorless to Yellow
Density 1.7000g/mL
Boiling Point 90.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4972 to 1.4992
Linear Formula (CH3)2CHI
Quantity 500 mL
Beilstein 01, 114
Specific Gravity 1.7
Merck Index 15, 5260
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with chloroform,ehter,alcohol and,benzene
Stabilizer copper
Formula Weight 169.99
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-859-3
RTECSNumber TZ4200000
TSCA TSCA

RUO – Research Use Only