missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopentane, tech. 90%

Catalog No. AAA1720522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1720522 100 g
AAA1720536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1720522 Supplier Thermo Scientific Chemicals Supplier No. A1720522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-81-3 | C5H11Br | 151.047 g/mol

Used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chemical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 107-81-3
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000160
InChI Key LGAJYTCRJPCZRJ-UHFFFAOYSA-N
Synonym pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
PubChem CID 7890
IUPAC Name 2-bromopentane
SMILES CCCC(C)Br

Specifications

Melting Point -96°C
Density 1.22
Boiling Point 117°C to 118°C
Flash Point 20°C (68°F)
Odor Characteristic
Refractive Index 1.4403
Quantity 100 g
UN Number UN2343
Beilstein 1718829
Solubility Information Not miscible or difficult to mix in water. Soluble in Dichloromethane,Ether,Ethyl Acetate,Hexane and Methanol
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Bromopentane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOPENTANE
EINECSNumber 203-521-3
RTECSNumber RZ9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only