missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)cyclohexane, 99%

Catalog No. AAL0946614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0946614 25 g
AAL0946603 1 g
AAL0946606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0946614 Supplier Thermo Scientific Chemicals Supplier No. L0946614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1647-26-3 | C8H15Br | 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1647-26-3
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.11
MDL Number MFCD00019398
InChI Key JRQAAYVLPPGEHT-UHFFFAOYSA-N
Synonym 2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane
PubChem CID 15440
IUPAC Name 2-bromoethylcyclohexane
SMILES BrCCC1CCCCC1

Specifications

Density 1.221
Boiling Point 70°C to 71°C (6 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.49
Quantity 25 g
Beilstein 1919816
Formula Weight 191.12
Percent Purity 99%
Chemical Name or Material (2-Bromoethyl)cyclohexane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-712-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only