missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutane, 99+%

Catalog No. AC106785000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106785000 500 mL Glass bottle
AC106781000 100 mL Glass bottle
AC106780025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106785000 Supplier Thermo Scientific Chemicals Supplier No. 106785000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-76-2 | C4H9Br | 137.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-76-2
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000156
InChI Key UPSXAPQYNGXVBF-UHFFFAOYSA-N
Synonym sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
PubChem CID 6554
IUPAC Name 2-bromobutane
SMILES CCC(C)Br

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -112°C
Density 1.2500g/mL
Boiling Point 91°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4359 to 1.4379
Linear Formula C2H5CH(Br)CH3
Quantity 500 mL
Beilstein 01,119
Merck Index 15,1556
Specific Gravity 1.25
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 137.02
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Bromobutane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 201-140-7
RTECSNumber EJ6228000
TSCA TSCA

RUO – Research Use Only