missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromohexane (mixture of diastereoisomers) 98.0+%, TCI America™

Catalog No. D22985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22985G 5 g
D229825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22985G Supplier TCI America Supplier No. D22985G

Chemical Identifiers

CAS 24774-58-1
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00039177
InChI Key MQYLGFBWOZXHHF-UHFFFAOYSA-N
Synonym hexane, 2,5-dibromo, dibromohexane, 2,5-dibromohexane, erythro + threo, 2.5-dibromhexan, acmc-1aqpr, 2,5-bis bromanyl hexane, mixture of diastereoisomers
PubChem CID 91232
IUPAC Name 2,5-dibromohexane
SMILES CC(CCC(C)Br)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 243.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromohexane (mixture of diastereoisomers)