missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobiphenyl 97.0+%, TCI America™

Catalog No. D34505G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34505G Supplier TCI America Supplier No. D34505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37847-52-2
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.193
MDL Number MFCD00042515
InChI Key JVHAJKHGPDDEEU-UHFFFAOYSA-N
PubChem CID 169974
IUPAC Name 2,4-difluoro-1-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C=C(C=C2)F)F

Specifications

Melting Point 63°C
Quantity 5 g
UN Number 3152
Formula Weight 190.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Difluorobiphenyl